missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MKK7/MEK7 Rabbit anti-Human, Mouse, Rat, Clone: 1T3M6, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £425.00
Specifications
| Antigen | MKK7/MEK7 |
|---|---|
| Clone | 1T3M6 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18388826
|
Novus Biologicals
NBP3-15456-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18301316
|
Novus Biologicals
NBP3-15456-100UL |
100 μg |
£425.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MKK7/MEK7 Monoclonal antibody specifically detects MKK7/MEK7 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| MKK7/MEK7 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 5609 | |
| IgG | |
| Affinity purified |
| 1T3M6 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| c-Jun N-terminal kinase kinase 2, dual specificity mitogen-activated protein kinase kinase 7, EC 2.7.12.2, JNK kinase 2, JNK-activating kinase 2, JNKK 2, JNKK2, MAP kinase kinase 7, MAPK/ERK kinase 7, MAPKK7, MEK 7, MEK7, mitogen-activated protein kinase kinase 7, MKK7MAPKK 7, PRKMK7 | |
| A synthetic peptide corresponding to a sequence within amino acids 320-419 of human MKK7/MEK7 (O14733). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title