missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MKK7/MEK7 Rabbit anti-Human, Mouse, Rat, Clone: 1T3M6, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-15456-100UL
This item is not returnable.
View return policy
Description
MKK7/MEK7 Monoclonal antibody specifically detects MKK7/MEK7 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| MKK7/MEK7 | |
| Monoclonal | |
| Unconjugated | |
| c-Jun N-terminal kinase kinase 2, dual specificity mitogen-activated protein kinase kinase 7, EC 2.7.12.2, JNK kinase 2, JNK-activating kinase 2, JNKK 2, JNKK2, MAP kinase kinase 7, MAPK/ERK kinase 7, MAPKK7, MEK 7, MEK7, mitogen-activated protein kinase kinase 7, MKK7MAPKK 7, PRKMK7 | |
| A synthetic peptide corresponding to a sequence within amino acids 320-419 of human MKK7/MEK7 (O14733). | |
| 100 μg | |
| Protein Kinase | |
| 5609 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 1T3M6 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction