missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MKK7/MEK7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-55001
This item is not returnable.
View return policy
Description
MKK7/MEK7 Polyclonal specifically detects MKK7/MEK7 in Human samples. It is validated for Western Blot.
Specifications
| MKK7/MEK7 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| c-Jun N-terminal kinase kinase 2, dual specificity mitogen-activated protein kinase kinase 7, EC 2.7.12.2, JNK kinase 2, JNK-activating kinase 2, JNKK 2, JNKK2, MAP kinase kinase 7, MAPK/ERK kinase 7, MAPKK7, MEK 7, MEK7, mitogen-activated protein kinase kinase 7, MKK7MAPKK 7, PRKMK7 | |
| Rabbit | |
| 47 kDa | |
| 100 μL | |
| Protein Kinase | |
| 5609 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Sheep, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| O14733 | |
| MAP2K7 | |
| Synthetic peptides corresponding to MAP2K7(mitogen-activated protein kinase kinase 7) The peptide sequence was selected from the middle region of MAP2K7. Peptide sequence ERIDPPDPTKPDYDIRADVWSLGISLVELATGQFPYKNCKTDFEVLTKVL. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction