missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MKK6/MEK6, p Ser207 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£203.00 - £324.00
Specifications
| Antigen | MKK6/MEK6 (p Ser207) |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:50-1:100 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18257764
|
Novus Biologicals
NB100-82050-0.025ML |
0.025 mL |
£203.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18127279
|
Novus Biologicals
NB100-82050 |
0.05 mL |
£324.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MKK6/MEK6 Polyclonal specifically detects MKK6/MEK6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| MKK6/MEK6 (p Ser207) | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:50-1:100 | |
| Polyclonal | |
| Rabbit | |
| Angiogenesis, Apoptosis, Cancer, Cell Cycle and Replication, Hypoxia, Phospho Specific | |
| P52564 | |
| 5608 | |
| IgG | |
| Affinity Purified |
| 1.0 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Rat, Human, Mouse | |
| MAP kinase kinase 6, MAPK/ERK kinase 6, MAPKK6dual specificity mitogen-activated protein kinase kinase 6, MEK6MAPKK 6, mitogen-activated protein kinase kinase 6, MKK6MEK 6, PRKMK6protein kinase, mitogen-activated, kinase 6 (MAP kinase kinase 6), SAPKK3EC 2.7.12.2 | |
| MAP2K6 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title