missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MKK6/MEK6, p Ser207 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-82050
This item is not returnable.
View return policy
Description
MKK6/MEK6 Polyclonal specifically detects MKK6/MEK6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| MKK6/MEK6 (p Ser207) | |
| Polyclonal | |
| Unconjugated | |
| PBS (without Mg2+ and Ca2+), pH 7.4, 150 mM NaCl, 50% glycerol with 0.02% Sodium Azide | |
| MAP kinase kinase 6, MAPK/ERK kinase 6, MAPKK6dual specificity mitogen-activated protein kinase kinase 6, MEK6MAPKK 6, mitogen-activated protein kinase kinase 6, MKK6MEK 6, PRKMK6protein kinase, mitogen-activated, kinase 6 (MAP kinase kinase 6), SAPKK3EC 2.7.12.2 | |
| Rabbit | |
| 38 kDa | |
| 0.05 mL | |
| Angiogenesis, Apoptosis, Cancer, Cell Cycle and Replication, Hypoxia, Phospho Specific | |
| 5608 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:50-1:100 | |
| P52564 | |
| MAP2K6 | |
| The antiserum was produced against synthesized phosphopeptide derived from human MKK6/MEK6 around the phosphorylation site of serine 207 (V-D-SP-V-A). | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Detects endogenous levels of MKK6/MEK6 only when phosphorylated at serine207. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction