missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Mixed Cellulose Ester Membranes Sterile Filter

Product Code. 11735285 Shop All Fisherbrand Products
Change view
Click to view available options
Pore Size:
0.2 μm
0.45 μm
Color:
Black
White
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Color unitSize
11735285 0.45 μm White Pack of 100
11745285 0.45 μm Black Pack of 100
11765285 0.2 μm White Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11735285

Please to purchase this item. Need a web account? Register with us today!

Fisherbrand™ Mixed Celluloses Esters Membranes Sterile Filters are composed of cellulose acetate and cellulose nitrate.

  • Widely used membranes in analytical and research applications
  • Naturally hydrophilic, biologically inert and offer thermal stability
  • Characterized by smoother and more uniform surface than pure nitrocellulose membrane which facilitates particles detection, and high retention level
  • Gridded membranes are used in many microbiological techniques include colony counting after incubation

Specifications

Bubble Point 2.23 bar
Color White
Material MCE
Pore Size 0.45 μm
Sterility Sterile
Flow Rate 60 mL/min./cm2
For Use With (Application) Microdialysis of DNA and Proteins, clarification of aqueous solutions during microbiological analysis, identification of Nucleic Acids
Gas Flow Rate 5 L/min./cm2
Grid Color Black
Hydrophobicity Hydrophilic
Quantity 100/Pk.
Type Gridded Disc
Wettability Yes
Diameter (Metric) 47 mm
Show More Show Less
compliance-icons
Product Identifier
  • X100 MBNE MCE WHIT/N 47MM .45µ ST Fisherbrand .
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P370+P378-In case of fire: Use to extinguish.
Supplemental information
  • MIXTURE LIST-Contain: nitrocellulose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.