missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIS RII/AMHR2 Rabbit anti-Human, Clone: 8Z8X4, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | MIS RII/AMHR2 |
|---|---|
| Clone | 8Z8X4 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18306722
|
Bio-Techne
NBP3-16706-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18342782
|
Bio-Techne
NBP3-16706-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MIS RII/AMHR2 Monoclonal antibody specifically detects MIS RII/AMHR2 in Human samples. It is validated for Western BlotSpecifications
| MIS RII/AMHR2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 269 | |
| IgG | |
| Affinity purified |
| 8Z8X4 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| AMH type II receptor, AMHREC 2.7.11.30, MISR2, MISRIIMIS type II receptor, MRII, Muellerian hormone type II receptor, Muellerian hormone type-2 receptor, Mullerian hormone receptor, type II, Mullerian inhibiting substance type II receptor | |
| A synthetic peptide corresponding to a sequence within amino acids 50-150 of human MIS RII/AMHR2 (Q16671). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title