missing translation for 'onlineSavingsMsg'
Learn More

MIS RII/AMHR2 Rabbit anti-Human, Clone: 8Z8X4, Novus Biologicals™

Product Code. 18306722
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18306722 20 μg 20µL
18342782 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18306722 Supplier Bio-Techne Supplier No. NBP31670620UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

MIS RII/AMHR2 Monoclonal antibody specifically detects MIS RII/AMHR2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIS RII/AMHR2
Applications Western Blot
Classification Monoclonal
Clone 8Z8X4
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias AMH type II receptor, AMHREC 2.7.11.30, MISR2, MISRIIMIS type II receptor, MRII, Muellerian hormone type II receptor, Muellerian hormone type-2 receptor, Mullerian hormone receptor, type II, Mullerian inhibiting substance type II receptor
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 50-150 of human MIS RII/AMHR2 (Q16671).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 269
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.