missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Mirin, 95%

Product Code. p-7057872
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 15478739

Brand: Thermo Scientific Chemicals J67462.#0

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 299953-00-7 | C10H8N2O2S, C10H8N2O2S | 220.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 299953-00-7
Molecular Formula C10H8N2O2S
Synonym cis-5-(4-Hydroxybenzylidene)-2-imino-4-thiazolidinone;

Specifications

Quantity 5 mg
Description Orange
Content And Storage -30°C to -10°C
Formula Weight 220.25g/mol
Sensitivity Moisture Sensitive
Form Solid
InChI Key YBHQCJILTOVLHD-UHFFFAOYSA-N
SMILES NC1=NC(=O)C(S1)=CC1=CC=C(O)C=C1
IUPAC Name 2-amino-5-[(4-hydroxyphenyl)methylidene]-4,5-dihydro-1,3-thiazol-4-one
Molecular Weight (g/mol) 220.25
Assay 95%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.