missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Mirin, 95%

Product Code. 15478739
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15478739 5 mg 5mg
15498739 50 mg 50mg
15488739 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15478739 Supplier Thermo Scientific Chemicals Supplier No. J67462.#0
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 299953-00-7 | C10H8N2O2S, C10H8N2O2S | 220.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 299953-00-7
Molecular Formula C10H8N2O2S
Synonym cis-5-(4-Hydroxybenzylidene)-2-imino-4-thiazolidinone;

Specifications

Quantity 5 mg
Description Orange
Content And Storage -30°C to -10°C
Formula Weight 220.25g/mol
Sensitivity Moisture Sensitive
Form Solid
InChI Key YBHQCJILTOVLHD-UHFFFAOYSA-N
SMILES NC1=NC(=O)C(S1)=CC1=CC=C(O)C=C1
IUPAC Name 2-amino-5-[(4-hydroxyphenyl)methylidene]-4,5-dihydro-1,3-thiazol-4-one
Molecular Weight (g/mol) 220.25
Assay 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.