Learn More
Invitrogen™ MIP-2/CXCL2 Mouse ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EMCXCL2X5
Additional Details : Weight : 0.33800kg
Description
The Mouse MIP-2 (CXCL2) ELISA quantitates Ms MIP-2 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms MIP-2. Principle of the method The Mouse MIP-2 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Macrophage inflammatory protein 2 is a chemokine involved in the pulmonary inflammatory response. MIP-2 is produced by activated monocytes and neutrophils and expressed at sites of inflammation. In vitro, it suppresses hematopoietic progenitor cell proliferation.Specifications
P10889 | |
10 pg/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
5 x 96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
20310 | |
<12% | |
SAV-HRP, Adhesive Plate Covers, Pre-coated 96 well plates, Standard, Biotinylated Detection Antibody, Stop Solution, Chromogen, Assay Diluent Concentrate, Wash Buffer | |
5 x 96 Tests | |
Immunology | |
2°C to 8°C | |
Mouse | |
4 hr. 45 min. |
2.74 pg/mL to 2000 pg/mL | |
10 pg/mL to 2000 pg/mL | |
Biotin | |
Chemokine & Receptors | |
This ELISA kit shows no cross-reactivity with any of the cytokines tested: Mouse CD30, L CD30, T CD40, CRG-2, CTACK, CXCL16, Eotaxin , Eotaxin-2, Fas Ligand, Fractalkine, GCSF, GM-CFS, IFN- gamma, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-3 Rb, IL- 4, IL-5, IL-6, IL-9, IL-10, IL-12 p40/p70, IL-12 p70, IL-13, IL-17, KC, Leptin R, LEPTIN(OB), LIX, L-Selectin, Lymphotactin, MCP-1, MCP-5, M-CSF, MIG, MIP-1 alpha, MIP-1 gamma, MIP-3 beta, MIP-3 alpha, PF-4, P-Selectin, RANTES, SCF, SDF-1 alpha, TARC, TCA-3, TECK, TIMP-1, TNF-alpha, TNF RI, TNF RII, TPO, VCAM-1, VEGF. | |
Mouse | |
Colorimetric Microplate Reader | |
CINC-2 A,GROb,Gro2,MIP-2,MIP-2 A,Mgsa-b,Mip2,Scyb,Scyb2 | |
<10% | |
HRP | |
RUO | |
Plasma, 50 μ;L; Serum, 50 μ;L; Supernatant, 100 μ;L | |
CINC-2 A, GROb, Gro2, MIP-2, MIP-2 A, Mgsa-b, Mip2, Scyb, Scyb2 | |
1 hr. 20 min. |