missing translation for 'onlineSavingsMsg'
Learn More

Minocycline hydrochloride, 890-950μg/mg, Thermo Scientific™

£150.65 - £207.00

Chemical Identifiers

CAS 13614-98-7
Molecular Formula C23H28ClN3O7
Molecular Weight (g/mol) 493.94
MDL Number MFCD00083669
InChI Key KDLQIOPKJDNQIM-WUURTAMISA-N
PubChem CID 54685925
IUPAC Name hydrogen (2Z,4S,4aS,5aR,12aS)-2-[amino(hydroxy)methylidene]-4,7-bis(dimethylamino)-10,11,12a-trihydroxy-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione chloride
SMILES [H+].[Cl-].CN(C)[C@H]1[C@@H]2C[C@@H]3CC4=C(C=CC(O)=C4C(O)=C3C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O)N(C)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15427789
SDS View Documents
Thermo Scientific Alfa Aesar
J66429.ME
500mg
ONLINE EXCLUSIVE PRICE
£167.00
£150.65 / 500mg
Save £16.35
9% Off
View alternative products
Save Up to 
»
Estimated Shipment: 30-09-2026
Add to basket
 
 Restricted Item
15417789
SDS View Documents
Thermo Scientific Alfa Aesar
J66429.03
1g
£207.00
1g
View alternative products
Save Up to 
»
Estimated Shipment: 30-09-2026
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • A broad spectrum antibiotic commonly used against bacteria which cause upper respiratory tract infections
Specifications

Chemical Identifiers

13614-98-7
493.94
KDLQIOPKJDNQIM-WUURTAMISA-N
hydrogen (2Z,4S,4aS,5aR,12aS)-2-[amino(hydroxy)methylidene]-4,7-bis(dimethylamino)-10,11,12a-trihydroxy-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione chloride
C23H28ClN3O7
MFCD00083669
54685925
[H+].[Cl-].CN(C)[C@H]1[C@@H]2C[C@@H]3CC4=C(C=CC(O)=C4C(O)=C3C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O)N(C)C

Specifications

13614-98-7
>110°C (230°F)
MFCD00083669
14,6202
KDLQIOPKJDNQIM-WUURTAMISA-N
hydrogen (2Z,4S,4aS,5aR,12aS)-2-[amino(hydroxy)methylidene]-4,7-bis(dimethylamino)-10,11,12a-trihydroxy-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione chloride
54685925
Minocycline hydrochloride, 890 to 950μg/mg
Yellow
C23H28ClN3O7
813°C
Freely soluble in water
[H+].[Cl-].CN(C)[C@H]1[C@@H]2C[C@@H]3CC4=C(C=CC(O)=C4C(O)=C3C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O)N(C)C
493.94
493.94
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 237-099-7

missing translation for 'rtecsNumber' : QI7630500

missing translation for 'tsca' : No

missing translation for 'storageNote' : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.