missing translation for 'onlineSavingsMsg'
Learn More

Minocycline hydrochloride, 890-950μg/mg, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J66429.03

Quantity 1g

 View more versions of this product

Product Code. 15417789

  • £207.00 / 1g
 Restricted Item
Estimated Shipment: 30-09-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • A broad spectrum antibiotic commonly used against bacteria which cause upper respiratory tract infections
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

13614-98-7
493.94
KDLQIOPKJDNQIM-WUURTAMISA-N
hydrogen (2Z,4S,4aS,5aR,12aS)-2-[amino(hydroxy)methylidene]-4,7-bis(dimethylamino)-10,11,12a-trihydroxy-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione chloride
C23H28ClN3O7
MFCD00083669
54685925
[H+].[Cl-].CN(C)[C@H]1[C@@H]2C[C@@H]3CC4=C(C=CC(O)=C4C(O)=C3C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O)N(C)C

Specifications

13614-98-7
1g
C23H28ClN3O7
813°C
Freely soluble in water
[H+].[Cl-].CN(C)[C@H]1[C@@H]2C[C@@H]3CC4=C(C=CC(O)=C4C(O)=C3C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O)N(C)C
493.94
493.94
Yellow
>110°C (230°F)
MFCD00083669
14,6202
KDLQIOPKJDNQIM-WUURTAMISA-N
hydrogen (2Z,4S,4aS,5aR,12aS)-2-[amino(hydroxy)methylidene]-4,7-bis(dimethylamino)-10,11,12a-trihydroxy-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione chloride
54685925
Minocycline hydrochloride, 890 to 950μg/mg
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.