missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mineralocorticoid R/NR3C2 Antibody (H10E4C9F) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
£169.00
Specifications
| Antigen | Mineralocorticoid R/NR3C2 |
|---|---|
| Clone | H10E4C9F |
| Concentration | 1.4 mg/mL |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:20, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100 - 1:200, Immunohistochemistry-Paraffin 1:100, Blocking/Neutralizing 50 - 200 molar excess |
| Applications | Western Blot |
Description
Mineralocorticoid R/NR3C2 Monoclonal specifically detects Mineralocorticoid R/NR3C2 in Human, Mouse, Rat, Chicken, Rabbit, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Block/Neutralize.Specifications
| Mineralocorticoid R/NR3C2 | |
| 1.4 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| FLJ41052, MGC133092, mineralocorticoid receptor 1, mineralocorticoid receptor delta, MLRmineralocorticoid receptor, MRMCRaldosterone receptor, NR3C2VIT, Nuclear receptor subfamily 3 group C member 2, nuclear receptor subfamily 3, group C, member 2 | |
| NR3C2 | |
| IgG1 | |
| Protein A purified | |
| Detects mineralocorticoid receptor (MR). |
| H10E4C9F | |
| Western Blot 1:1000, Flow Cytometry 1:20, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100 - 1:200, Immunohistochemistry-Paraffin 1:100, Blocking/Neutralizing 50 - 200 molar excess | |
| Monoclonal | |
| Purified | |
| RUO | |
| P08235 | |
| 4306 | |
| Aldosterone-3 | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title