missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mineralocorticoid R/NR3C2 Antibody (H10E4C9F) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB300-562-0.025ml
This item is not returnable.
View return policy
Description
Mineralocorticoid R/NR3C2 Monoclonal specifically detects Mineralocorticoid R/NR3C2 in Human, Mouse, Rat, Chicken, Rabbit, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Block/Neutralize.
Specifications
| Mineralocorticoid R/NR3C2 | |
| Monoclonal | |
| 1.4 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:20, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100 - 1:200, Immunohistochemistry-Paraffin 1:100, Blocking/Neutralizing 50 - 200 molar excess | |
| FLJ41052, MGC133092, mineralocorticoid receptor 1, mineralocorticoid receptor delta, MLRmineralocorticoid receptor, MRMCRaldosterone receptor, NR3C2VIT, Nuclear receptor subfamily 3 group C member 2, nuclear receptor subfamily 3, group C, member 2 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 4306 | |
| Human | |
| Purified |
| Western Blot | |
| H10E4C9F | |
| Unconjugated | |
| P08235 | |
| NR3C2 | |
| Aldosterone-3 | |
| 0.025 mL | |
| Primary | |
| Detects mineralocorticoid receptor (MR). | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction