missing translation for 'onlineSavingsMsg'
Learn More

Merck Millex™ Syringe Filter, PES, 0.22 μm
BIOPHARMA

Product Code. 11755223 Shop All Merck Products
Change view
Click to view available options
Diameter (Metric) Filter:
13 mm
33 mm
Quantity:
100/Pk.
1000/Pk.
250/Pk.
50/Pk.
Unit Size:
Pack of 100
Pack of 1000
Pack of 250
Pack of 50
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Filter Quantity unitSize
11755223 13 mm 1000/Pk. Pack of 1000
11790453 13 mm 100/Pk. Pack of 100
10509952 33 mm 250/Pk. Pack of 250
10163282 33 mm 1000/Pk. Pack of 1000
10747984 33 mm 50/Pk. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11755223 Supplier Merck Supplier No. SLGPX13NK
Add to basket
Add to basket
This item is not returnable. View return policy

Millex™ PES Syringe Filter, 0.22 μm for removing particulates, reducing signal-to-noise ratios and maintaining flat baselines.

Overview of Non-Sterile Millex™ Syringe Filters with PES Membrane:

  • Non-sterile Millex™ syringe filters with Millipore Express™ PLUS polyethersulfone (PES) membrane provide fast filtration of aqueous and mild organic solutions.
  • Low protein binding to minimize interaction with your sample and maximize recovery.
  • Polypropylene housing, offering a low level of extractables and broad chemical compatibility.

Applications:

  • Buffer Filtration, Clarification of Aqueous Solutions
TRUSTED_SUSTAINABILITY

Specifications

Color Code Green
Connectors Male Luer Slip
Disposable Single-use
Filter Type Syringe Filter
For Use With (Equipment) Syringe
Material Polypropylene
Material (Membrane) PES
Pore Size 0.22 μm
Sample Volume 10 mL
Sterility Non-sterile
For Use With (Application) Analytical sample prep
Housing Material Polypropylene
Hydrophobicity Hydrophilic
Inlet Style Male Luer Slip
Outlet Style Male Luer Slip
Quantity 1000/Pk.
Type Syringe Filter
Volume (Metric) <10 mL
Diameter (Metric) 13 mm
Diameter (Metric) Filter 13 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.