missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Overview of Non-Sterile Millex™ Syringe Filters with PES Membrane:
- Non-sterile Millex™ syringe filters with Millipore Express™ PLUS polyethersulfone (PES) membrane provide fast filtration of aqueous and mild organic solutions.
- Low protein binding to minimize interaction with your sample and maximize recovery.
- Polypropylene housing, offering a low level of extractables and broad chemical compatibility.
Applications:
- Buffer Filtration, Clarification of Aqueous Solutions
Specifications
Specifications
| Connectors | Male Luer Slip |
| Disposable | Single-use |
| Filter Type | Syringe Filter |
| For Use With (Equipment) | Syringe |
| Material | Polypropylene |
| Material (Membrane) | PES |
| Pore Size | 0.22 μm |
| Sample Volume | 100 mL |
| Sterility | Non-sterile |
| Filtration Area | 4.5 cm2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
