missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Millex™-GP Syringe Filter, PES, 0.45 μm
Ideal for preparing 10–100mL samples prior to chromatography or other instrumental analyses
£224.00 - £1680.00
Specifications
Disposable | Single-use |
---|---|
Pore Size | 0.45 μm |
Sterility | Non-sterile |
For Use With (Application) | Analytical sample prep |
Housing Material | PP |
Product Code | Brand | Diameter (Metric) Filter | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Filter | Quantity | Price | Quantity & Availability | ||||
10664923
|
Merck
SLHP033NB |
33 mm | 250Pack |
£650.00
Pack of 250 |
Estimated Shipment: 21-06-2024
Log in to see stock. |
||||
11700463
|
Merck
SLHPX13NL |
13 mm | 100Pack |
£326.00
Pack of 100 |
Estimated Shipment: 24-05-2024
Log in to see stock. |
||||
10509582
|
Merck
SLHP033NS |
33 mm | 50Pack |
£224.00
Pack of 50 |
Estimated Shipment: 18-11-2024
Log in to see stock. |
||||
11765223
|
Merck
SLHPX13NK |
13 mm | 1000Pack |
£1680.00
Pack of 1000 |
Estimated Shipment: 25-06-2024
Log in to see stock. |
||||
10665113
|
Merck
SLHP033NK |
33 mm | 1000Pack |
£1670.00
Pack of 1000 |
Estimated Shipment: 25-06-2024
Log in to see stock. |
||||
Description
Ideal for preparing 10–100mL samples prior to chromatography or other instrumental analyses
Overview of Non-Sterile Millex™ Syringe Filters with PES Membrane:- Non-sterile Millex™ syringe filters with Millipore Express™ PLUS polyethersulfone (PES) membrane provide fast filtration of aqueous and mild organic solutions.
- Available in 0.22 μm and 0.45 μm pore sizes and two diameters to suit your application needs
- Low protein binding to minimize interaction with your sample and maximize recovery
- Polypropylene housing, offering a low level of extractables and broad chemical compatibility
Applications: Buffer Filtration, Clarification of Aqueous Solutions
Specifications
Single-use | |
Non-sterile | |
PP |
0.45 μm | |
Analytical sample prep | |
Syringe Filter |