missing translation for 'onlineSavingsMsg'
Learn More

Merck Millex™-GP Syringe Filter, PES, 0.45 μm

Ideal for preparing 10–100mL samples prior to chromatography or other instrumental analyses

Brand:  Merck SLHPX13NL

287.13 GBP valid until 2024-12-31
Use promo code "23269" to get your promotional price.



Code : 71

Additional Details : Weight : 0.16168kg

 View more versions of this product

Product Code. 11700463

  • £326.00 / Pack of 100
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

Ideal for preparing 10–100mL samples prior to chromatography or other instrumental analyses

Overview of Non-Sterile Millex™ Syringe Filters with PES Membrane:
  • Non-sterile Millex™ syringe filters with Millipore Express™ PLUS polyethersulfone (PES) membrane provide fast filtration of aqueous and mild organic solutions.
  • Available in 0.22 μm and 0.45 μm pore sizes and two diameters to suit your application needs
  • Low protein binding to minimize interaction with your sample and maximize recovery
  • Polypropylene housing, offering a low level of extractables and broad chemical compatibility

Applications: Buffer Filtration, Clarification of Aqueous Solutions

Specifications

Specifications

Green
Hydrophilic PES
Non-sterile
PP
Male Luer slip
Syringe Filter
13 mm
Single-use
0.45 μm
Analytical sample prep
male luer slip
100Pack
<10 mL
13 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers