Learn More
Honeywell™ Millennia 2G Safety Glasses
Click to view available options
Lens Tint:
Clear
Gray
Silver
Unit Size:
Each
Description
- Softer, more flexible frame material for superior comfort against the head and ears when worn for all-day wear
- Covers the eyes and areas above the cheeks and brow to ensure that the workers are protected from particulates, dust and other impact risks
- A greater lens coverage wrap both peripherally and along the cheeks helps prevent particulate risk
- A “shelf-like” design element along the brow has been added to help to prevent dust from dropping in between the frame and entering the eyes
Specifications
Specifications
| Lens Coating | Anti-fog |
| Description | Dielectric |
| Certifications/Compliance (Europe) | EC Category PPE: 2, EC Certificate: C1570.1HON, EC Attestation: C1570.1HON, EN C1570.1HON |
| Lens Tint | Gray |
| For Use With (Application) | For outdoor use where eye protection is required against most commonly encountered hazards including impact protection, fogging, and sunlight glare. |
| Frame Color | Black |
| Lens Material | Polycarbonate |
| Frame Material | Nylon |
| UV Protection | UV400 |