missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifikationer
| Antigen | MIF |
|---|---|
| Concentration | 0.3 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Produktkod | Brand | Quantity | Pris | Kvantitet och tillgänglighet | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkod | Brand | Quantity | Pris | Kvantitet och tillgänglighet | |||||
|
16819954
|
Proteintech
20415-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16809954
|
Proteintech
20415-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivning
MIF (macrophage migration inhibitory factor) was one of the first cytokine activities to be discovered and was initially described as a T cell-derived factor that inhibits the random migration of macrophages (Weiser 1989). Recently, MIF was rediscovered as a pituitary hormone that act as the counterregulatory hormone for glucocorticoid action within the immune system (Bernhagen 1993, Mitchell 1995). MIF was released from macrophages and T cells in response to physiological concentrations of glucocorticoids. The secreted MIF counter-regulates the immunosuppressive effects of steroids on immune cell activation and cytokine production (Bucala 1998). MIF plays a critical role in the host control of inflammation and immunity.Specifikationer
| MIF | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| P14174, P30904, P34884 | |
| 17319, 4282, 81683 | |
| MIF Fusion Protein Ag14058 | |
| Primary | |
| -20°C | |
| MIF |
| 0.3 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| GIF, GLIF, L dopachrome isomerase, L dopachrome tautomerase, MIF, MMIF, Phenylpyruvate tautomerase | |
| Mif | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel