missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20415-1-AP-20UL
This item is not returnable.
View return policy
Description
MIF (macrophage migration inhibitory factor) was one of the first cytokine activities to be discovered and was initially described as a T cell-derived factor that inhibits the random migration of macrophages (Weiser 1989). Recently, MIF was rediscovered as a pituitary hormone that act as the counterregulatory hormone for glucocorticoid action within the immune system (Bernhagen 1993, Mitchell 1995). MIF was released from macrophages and T cells in response to physiological concentrations of glucocorticoids. The secreted MIF counter-regulates the immunosuppressive effects of steroids on immune cell activation and cytokine production (Bucala 1998). MIF plays a critical role in the host control of inflammation and immunity.Specifications
| MIF | |
| Polyclonal | |
| Unconjugated | |
| MIF | |
| GIF, GLIF, L dopachrome isomerase, L dopachrome tautomerase, MIF, MMIF, Phenylpyruvate tautomerase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17319, 4282, 81683 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P14174, P30904, P34884 | |
| Mif | |
| MIF Fusion Protein Ag14058 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction