missing translation for 'onlineSavingsMsg'
Learn More

Midecamycin, 900μg/mg, Thermo Scientific™

A broad spectrum antibiotic

£130.00 - £205.00

Chemical Identifiers

CAS 35457-80-8
Molecular Formula C41H67NO15
Molecular Weight (g/mol) 813.979
MDL Number MFCD00865000
InChI Key DMUAPQTXSSNEDD-RBATUFKESA-N
PubChem CID 124225192
IUPAC Name [(4R,5R,6S,7R,9R,10R,11E,13Z,16R)-6-[(2S,3R,4R,5S,6R)-4-(dimethylamino)-3-hydroxy-5-[(2S,4R,5S,6S)-4-hydroxy-4,6-dimethyl-5-propanoyloxyoxan-2-yl]oxy-6-methyloxan-2-yl]oxy-10-hydroxy-5-methoxy-9,16-dimethyl-2-oxo-7-(2-oxoethyl)-1-oxacyclohexadeca-11,13-di
SMILES CCC(=O)OC1CC(=O)OC(CC=CC=CC(C(CC(C(C1OC)OC2C(C(C(C(O2)C)OC3CC(C(C(O3)C)OC(=O)CC)(C)O)N(C)C)O)CC=O)C)O)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15487249
SDS View Documents
Thermo Scientific Alfa Aesar
J66046.ME
500mg
£130.00
500mg
View alternative products
Save Up to 
»
Estimated Shipment: 08-10-2026
Add to basket
 
 Restricted Item
15477249
SDS View Documents
Thermo Scientific Alfa Aesar
J66046.03
1g
£205.00
1g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

35457-80-8
813.979
DMUAPQTXSSNEDD-RBATUFKESA-N
[(4R,5R,6S,7R,9R,10R,11E,13Z,16R)-6-[(2S,3R,4R,5S,6R)-4-(dimethylamino)-3-hydroxy-5-[(2S,4R,5S,6S)-4-hydroxy-4,6-dimethyl-5-propanoyloxyoxan-2-yl]oxy-6-methyloxan-2-yl]oxy-10-hydroxy-5-methoxy-9,16-dimethyl-2-oxo-7-(2-oxoethyl)-1-oxacyclohexadeca-11,13-di
C41H67NO15
MFCD00865000
124225192
CCC(=O)OC1CC(=O)OC(CC=CC=CC(C(CC(C(C1OC)OC2C(C(C(C(O2)C)OC3CC(C(C(O3)C)OC(=O)CC)(C)O)N(C)C)O)CC=O)C)O)C

Specifications

35457-80-8
>110°C (230°F)
MFCD00865000
Soluble in methanol or ethanol; Insoluble in water
CCC(=O)OC1CC(=O)OC(CC=CC=CC(C(CC(C(C1OC)OC2C(C(C(C(O2)C)OC3CC(C(C(O3)C)OC(=O)CC)(C)O)N(C)C)O)CC=O)C)O)C
813.979
813.97
White
C41H67NO15
874°C
DMUAPQTXSSNEDD-RBATUFKESA-N
[(4R,5R,6S,7R,9R,10R,11E,13Z,16R)-6-[(2S,3R,4R,5S,6R)-4-(dimethylamino)-3-hydroxy-5-[(2S,4R,5S,6S)-4-hydroxy-4,6-dimethyl-5-propanoyloxyoxan-2-yl]oxy-6-methyloxan-2-yl]oxy-10-hydroxy-5-methoxy-9,16-dimethyl-2-oxo-7-(2-oxoethyl)-1-oxacyclohexadeca-11,13-di
124225192
Midecamycin, 900μg/mg
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 252-578-0

missing translation for 'rtecsNumber' : OH4730600

missing translation for 'tsca' : No

missing translation for 'storageNote' : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.