missing translation for 'onlineSavingsMsg'
Learn More

Midecamycin, 900μg/mg

Product Code. 15487249
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15487249 500 mg 500mg
15477249 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15487249 Supplier Thermo Scientific Alfa Aesar Supplier No. J66046.ME
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A broad spectrum antibiotic | CAS: 35457-80-8 | C41H67NO15 | 813.979 g/mol

A broad spectrum antibiotic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35457-80-8
Molecular Formula C41H67NO15
Molecular Weight (g/mol) 813.979
MDL Number MFCD00865000
InChI Key DMUAPQTXSSNEDD-RBATUFKESA-N
PubChem CID 124225192
IUPAC Name [(4R,5R,6S,7R,9R,10R,11E,13Z,16R)-6-[(2S,3R,4R,5S,6R)-4-(dimethylamino)-3-hydroxy-5-[(2S,4R,5S,6S)-4-hydroxy-4,6-dimethyl-5-propanoyloxyoxan-2-yl]oxy-6-methyloxan-2-yl]oxy-10-hydroxy-5-methoxy-9,16-dimethyl-2-oxo-7-(2-oxoethyl)-1-oxacyclohexadeca-11,13-di
SMILES CCC(=O)OC1CC(=O)OC(CC=CC=CC(C(CC(C(C1OC)OC2C(C(C(C(O2)C)OC3CC(C(C(O3)C)OC(=O)CC)(C)O)N(C)C)O)CC=O)C)O)C

Specifications

Type Midecamycin
Physical Form Powder
Melting Point 155°C to 156°C
Color White
Quantity 500 mg
Flash Point >110°C (230°F)
Boiling Point 874°C
Solubility Information Soluble in methanol or ethanol; Insoluble in water
Formula Weight 813.97
Chemical Name or Material Midecamycin, 900μg/mg

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.