missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Motic™ Phase Ring for AE2000
Increases image contrast. Motic™ Phase Ring for AE2000 eliminates the phase difference between the background and scattered light rays.
£32.70 - £55.10
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
11722358
|
Motic™
1101000201641 |
£33.85
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
||||||
11732358
|
Motic™
1101000201651 |
£32.70
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||||||
11840153
|
Motic™
1101001100391 |
PH2 |
£55.10
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
Description
- For use with AE2000 Binocular Inverted Microscopes
- Use Phase Ring Ph0 (4X) for working in phase contrast with the 4X objective
- Use Phase Ring Ph1 (10X, 20X, 40X) for working in phase contrast with the Ph10X, Ph20X, Ph40X objectives
Specifications
1 Ea. |