missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Motic™ Phase Ring for AE2000
Click to view available options
Unit Size:
Each
For Use With (Equipment):
Ph 4X objective
Ph10X, Ph20X, and Ph40X Objectives
Description
- For use with AE2000 Binocular Inverted Microscopes
- Use Phase Ring Ph0 (4X) for working in phase contrast with the 4X objective
- Use Phase Ring Ph1 (10X, 20X, 40X) for working in phase contrast with the Ph10X, Ph20X, Ph40X objectives
Specifications
Specifications
| Type | Phase Slider PH2 |
| Quantity | 1 Ea. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction