missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Microsart™ Research Mycoplasma Real-Time PCR Kit
For fast and reliable screening of cell culture supernatants. Microsart™ Research Mycoplasma Real-Time PCR Kit is used for direct detection of Mollicutes in cell culture, media components, and derived biologicals.
£575.00 - £1857.60
Specifications
Content And Storage | 2° to 8°C |
---|---|
Detection Method | qPCR |
Product Type | Assay test |
Target | 16s rRNA of Mycoplasma |
For Use With (Application) | Microbial Detection |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15378156
|
Sartorius
SMB95-1005 |
25 tests |
£575.00
25 tests |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15358176
|
Sartorius
SMB95-1006 |
100 tests |
£1857.60
100 tests |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Enables fast and reliable detection of Mycoplasma DNA in cell culture supernatants
- Detects 16 S rRNA gene of at least 110 Mycoplasma species
- Time: Three hours
- Sample can be added directly to PCR reaction tube
- TaqMan™ real-time qPCR based chemistry
- Sample volume: 200μL
Applications:
- Used with any Real-time PCR instrument which detects the dyes FAM, ROX
- Used in quality control analysis in microbiological and biopharmaceutical labs
Compliance
EP 2.6.7
Specifications
2° to 8°C | |
Assay test | |
Microbial Detection | |
2μL |
qPCR | |
16s rRNA of Mycoplasma | |
Lyophilized |