Learn More
Sartorius Stedim Biotech™ Microsart™ e.motion Membrane Filter Dispenser
Sartorius Stedim Biotech™ Microsart™ e.motion Membrane Filter Dispenser facilitates procedures by automatically dispensing membrane filters.
Brand:  Sartorius Stedim Biotech™ 16713----PS
Additional Details : Weight : 3.20000kg
 View more versions of this product
Product Code. 15963040
  • £2225.00 / Each
Estimated Shipment: 11-06-2024
to see stock.
Add to basket
Description
Description
Sartorius Stedim Biotech™ Microsart™ e.motion Membrane Filter Dispenser facilitates procedures by automatically dispensing membrane filters.
  • Facilitates microbiological testing procedures.
  • Small footprint saves precious laminar flow hood space.
  • Dispenses membrane filters from their sterile packaging fully automatically at the touch of a button, hands-free, or by foot switch.
Specifications
Specifications
2.6 kg
0.5 sec.
Environmental/water testing, food and beverage testing, pharma/biotech
2 V DC, 40VA (external power supply included)
233 mm × 172 mm × 230 mm
Membrane filters