Learn More
Sartorius Stedim Biotech™ Microsart™ Cellulose Nitrate Membrane Filter Units, Gridded, White/Black, 0.45μm
Sartorius Stedim Biotech™ Microsart™ Cellulose Nitrate Membrane Filter Units, Gridded, White/Black 0.45μm combine a funnel, a filter base and a gridded membrane filter in one unit.
Brand: Sartorius Stedim Biotech™ 16D01--25-06--BK
Additional Details : Weight : 0.72000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II
Description
- Ready-to-use, sterile filter units combine a funnel, a filter base, and a gridded membrane filter in one unit.
- Suitable for the detection and enumeration of microorganisms in pharmaceuticals, cosmetics, food, beverages, water, and other liquids.
- Suitable for use in pharma/biotech, food and beverage, environmental/water testing.
- Suitable for microbial enumeration.
- Sterilized by gamma irradiation.
Specifications
White/Black Grid | |
Polypropylene Funnel, ABS Base | |
0.45 μm | |
6 bags of 8 |
Funnel capacity: 250mL | |
Cellulose Nitrate | |
Sterile | |
47 mm |
Safety and Handling
- X48 Microsart Filter250,CN white-black,0.45µm
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable solid Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose