Learn More
Thermo Scientific™ Microplate Reader Verification Plates
Product Code. 32096685
Change view
Click to view available options
Product Type:
Lumiwell Luminometric Verification Plate (Lum)
Multifunctional Verification Plate (Abs+Fl)
Multiskan Verification Plate (Abs), 16 Wavelengths
Multiskan Verification Plate (Abs), 8 Wavelengths
Spectrophotometric Verification Plate (Abs)
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
32096685 Multifunctional Verification Plate (Abs+Fl) Each
11958995 Multiskan Verification Plate (Abs), 8 Wavelengths Each
17104669 Multiskan Verification Plate (Abs), 16 Wavelengths Each
12463458 Lumiwell Luminometric Verification Plate (Lum) Each
11560715 Spectrophotometric Verification Plate (Abs) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 32096685 Supplier Thermo Scientific™ Supplier No. N03394M2
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Thermo Scientific™ Microplate Reader Verification Plates are designed to be convenient solutions for verifying and documenting the optical performance of compatible Thermo Scientific™ microplate readers.
Thermo Scientific™ Microplate Reader Verification Plates are designed to be convenient solutions for verifying and documenting the optical performance of compatible Thermo Scientific™ microplate readers. The portfolio includes options for photometric, fluorometric, and luminometric performance verification, helping laboratories maintain confidence in measurement quality and instrument consistency over time.
Available verification plates
  • Multifunctional Verification Plate (N03394M2): Verifies photometric and fluorometric performance. Compatible with Varioskan™ LUX, Varioskan™ ALF, and Multiskan™ SkyHigh readers. Supported tests depend on the instrument model and may include photometric accuracy, precision, wavelength accuracy (verifies simultaneously fluorometric excitation wavelength accuracy) and stray light. With fluorometry, both top and bottom sensitivity and fluorescence emission wavelength accuracy can be verified.
  • Spectrophotometric Verification Plate (N03394): Verifies photometric accuracy, precision, linearity, wavelength accuracy, and stray light. Compatible with Varioskan™ LUX, Varioskan™ ALF, and Multiskan™ SkyHigh readers.
  • Multiskan Verification Plate, 8 wavelengths (24072800) or 16 wavelengths (24072805): Verifies photometric performance across 8 or 16 wavelengths. Compatible with Multiskan™ Ease (24072800) and Multiskan™ FC (24072800 or 24072805) readers.
  • Lumiwell Luminometric Verification Plate (2806460): Provides an independent Relative Light Unit reference for luminometric performance verification. Compatible with Varioskan™ LUX and Varioskan™ ALF readers.
The verification plates include certified optical reference materials appropriate for the supported detection technology. Photometric reference values are traceable to standards from the National Physical Laboratory (NPL, UK) and/or the National Institute of Standards and Technology (NIST, USA), as specified for each product. A Certificate of Calibration is supplied to support verification documentation.
The products include ready-to-use Thermo Scientific™ SkanIt™ Software sessions loading results automatically to Microsoft™ Excel™ data analysis template files to simplify measurement, result evaluation, and documentation.
Key features
  • Options for photometric, fluorometric, and luminometric verification
  • Supports routine performance verification and documentation
  • Certified reference values for reliable result evaluation
  • Designed for compatible Thermo Scientific™ microplate readers
  • Preconfigured SkanIt Software sessions and analysis files included
Includes:
  • Verification plate
  • Certificate of Calibration
  • Verification plate adapter, where required
  • USB drive with sessions, reference and data analysis template files
Protective carrying case
TRUSTED_SUSTAINABILITY
Specifications
Product Type Multifunctional Verification Plate (Abs+Fl)
For Use With (Equipment) Varioskan ALF , Varioskan LUX, Multiskan Skyhigh
Plate Type Verification Plate
Includes NPL/NIST traceable verification plate, calibration certificate, adapter, USB reference files, carrying case