missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Microglass Fiber Filters, Grade 261

Product Code. 11714083 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric):
25 mm
47 mm
70 mm
90 mm
110 mm
125 mm
150 mm
Unit Size:
Pack of 100
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
11714083 90 mm Pack of 100
11512083 25 mm Pack of 100
11704083 47 mm Pack of 100
11532083 70 mm Pack of 100
11724083 110 mm Pack of 100
11734083 125 mm Pack of 100
11744083 150 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 missing translation for 'options'
Product Code. 11714083
Only null left
Add to basket
Edge
Add to basket

Made with 100% borosilicate micro-glass fibers and are binder free. Fisherbrand™ Microglass Fiber Filters are particularly suitable for micro-filtration because of very fine fibers.

  • High loading capacity
  • Excellent levels of particles retention (pore size 0.7 to 2.7μm)
  • Chemical resistance to many solvents and reagents
  • Non-hygroscopic and biologically inert
  • HIgh permeability against the passing of air
  • Suitable for use at and up to 500°C
Applications: filtration of industrial water, ligand binding test, proteins filtration, radio-immuno tests, suspended sollids in water (EN872 norm), drinking water filtration, cells collection

Specifications

Basis Weight 52 g/m2
Filter Type Microfilter
Format Disc
Material Borosilicate Glass
Pore Size 1.2 μm
Filtration Time 100 sec.
For Use With (Application) Filtration of industrial water, ligand binding test, Proteins filtration, radio-immuno tests, suspended solids in water, drinking water filtration, cells collection
Grade 261
Surface Type Non-hygroscopic
Temperature (Metric) Max 500°C
Thickness (English) 0.01 in.
Thickness (Metric) 0.26 mm
Type Filter
Diameter (Metric) 90 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.