missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Microglass Fiber Filter Discs

Product Code. 11764083 Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (Metric):
47 mm
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
11764083 47 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11764083
Only null left
Add to basket
Edge
Add to basket

Made with 100% borosilicate micro-glass fibers. Fisherbrand™ Microglass Fiber Filter Discs are non-hygroscopic and biologically inert. They show chemical resistance to most solvents and reagents (except hydrofluoric acid).

  • Contains no binder
  • Suitable for use at and up to 500°C
  • Not affected by humidity
  • Suitable for micro-filtration due to fine fibers
  • High-loading capacity
  • High chemical purity
  • made with demineralized water
  • High permeability to air

Applications:

  • Filtration of industrial water, fresh water algae, drinking water, Proteins, samples for HPLC, solvents
  • Sediments content in petroleum products
  • Ligand binding test
  • Air pollution monitoring
  • Membranes pre-filters
  • Radio-immuno tests
  • Suspended solids in water, smokes
  • Liquid scintillation counting
  • Cells collection
  • Grease content in dairy products

Specifications

Basis Weight 75 g/m2
Filter Type Microfilter
Format Disc
Material Borosilicate Glass
Pore Size 0.7 μm
Filtration Time 310 sec.
For Use With (Application) Air pollution control, separation of soot before gas analyzers, support weighing in the balance analyzes and dryers
Grade 263
Quantity 100 Pack
Surface Type Non-hygroscopic
Temperature (Metric) Max 500°C
Thickness (English) 0.017 in.
Thickness (Metric) 0.45 mm
Type Filter
Diameter (Metric) 47 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.