Learn More
Ansell Microgard™ 2000 Standard White Coveralls with Hood and Boots
Product Code. 15536524 Shop All Ansell Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 30
Product Code. 15536524
Brand: Ansell WH20B0012206
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed to allow water vapor (perspiration) to escape from the suit yet will withstand saturation of liquid chemicals. Filter 100% of particulates down to 0.01 microns in size.
  • Coverall with 3-piece hood and boots with anti-slip soles
  • White bound seams
  • Elasticated hood, waist and wrists
  • Includes finger loops
  • 2-way front zipper with resealable storm flap
  • Anti-static treatment
TRUSTED_SUSTAINABILITY
Specifications
Product Type General Purpose Coveralls
Size 2X-Large
Gender Unisex
Material Microporous Polyethylene Laminate (Non-woven)
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel, Elastic
Closure Type Front Zipper with Storm Flap
Seam Type Bound
Sterility Non-sterile
Resistance Chemicals
Certifications/Compliance (Europe) Cat III Type 5-B, 6-B, EN1073-2, EN14126, EN1149-1, DIN 32781
Description Hood, boots, and finger loops
Model WH20-B-00-122
No. of Pockets None
Style Includes Hood, Boots, and Fingerloops