Learn More
Ansell Microgard™ 2000 Standard SOCO/CSI Coveralls with Hood
Product Code. 15566524 Shop All Ansell Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 40
Product Code. 15566524
Brand: Ansell WH20B0012804
Only null left
Add to basket
Add to basket
Designed to allow water vapor (perspiration) to escape from the suit yet will withstand saturation of liquid chemicals. Filter 100% of particulates down to 0.01 microns in size.
  • Self adhesive pockets can be positioned anywhere on the garment to secure equipment
  • Finger loops prevent sleeve movement when working above your head
  • Protection from biological agents in the highest performance class according to EN14126
  • Anti-static according to EN 1149-5
  • Elasticated hood, wrists, waist and ankles
  • 2-way front zipper with resealable storm flap
  • 2-piece hood with chinstrap
  • Dual finger loops
  • Includes 2 pockets
TRUSTED_SUSTAINABILITY
Specifications
Product Type General Purpose Coveralls
Size Large
Gender Unisex
Material Microporous Polyethylene Laminate (Non-woven)
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 2-panel with Chin Strap
Closure Type Front Zipper with Storm Flap
Seam Type Bound
Sterility Non-sterile
Resistance Chemicals
Certifications/Compliance (Europe) Cat III Type 5-B, 6-B, EN1073-2, EN14126, EN1149-1, DIN 32781
Description Hood with chinstrap, dual finger loops
Model WH20-B-00-128
No. of Pockets 2
Style Includes Hood with Chinstrap, Dual Fingerloops