missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) MicroFunnel™ ST Filters Funnel with Supor Membrane
Microfunnel ST filter funnels are available in gamma-irradiated, double-bagged packaging saving valuable time during entry into cleanrooms and hoods.
£214.00 - £385.00
Product Code | Brand | Pore Size | Capacity (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Capacity (Metric) | Price | Quantity & Availability | ||||
17174421
|
Cytiva (formerly Pall Lab)
4851 |
0.2 μm | 100 mL |
£385.00
Pack of 2 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
17184391
|
Cytiva (formerly Pall Lab)
4750 |
0.45 μm | 100 mL |
£325.00
Pack of 40 |
Estimated Shipment: 21-06-2024
Log in to see stock. |
||||
17194391
|
Cytiva (formerly Pall Lab)
4751 |
0.45 μm | 300 mL |
£214.00
Pack of 20 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
17194421
|
Cytiva (formerly Pall Lab)
4853 |
0.2 μm | 300 mL |
£263.00
Pack of 20 |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
Description
- For research use only
Specifications
Sterile | |
Supor (PES) |