Learn More
Ansell™ Microchem™ 4000 Coveralls with Hood
Product Code. 15586434 Shop All Ansell Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 10
Product Code. 15586434
Brand: Ansell™ GR40T0011103
Only null left
Add to basket
Add to basket
Provides an exceptional barrier against organic and inorganic chemicals and biological agents. Ansell Microchem™ 4000 Coveralls with Hood is constructed of a unique multi-layer fabric known for its lightwieght, yet robust textile feel.
  • Coverall with elasticated 2-piece hood and front entry double-zip system.
  • Elasticated double cuffs (knitted inner cuff), waist and ankles.
  • Permeation tested against over 180 chemicals, including chemical warfare agents
  • Textile-like inner improves wearer acceptance
  • Anti-static according to EN 1149-5
TRUSTED_SUSTAINABILITY
Specifications
Product Type Protective Coveralls
Size Medium
Gender Unisex
Material Multi-layer Barrier Laminate Fabric
Color Green
Disposable Limited-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Double, Elastic
Neck Style Hooded
Hood Type 2-panel, Elastic
Closure Type Double Zipper Front Entry
Seam Type Taped, Welded
Resistance Chemicals
Certifications/Compliance Type 3/4 EN14605, Type 5 EN ISO 13982-1, EN14126, EN1073-2, EN1149-5
No. of Pockets None
Style Includes Hood