Learn More
Eppendorf™ Micro Fusion Chamber
Easy to use. Integrated cuvette chamber and extremely high transformation rates
Brand:  Eppendorf™ 4308031000
 View more versions of this product
Product Code. 15204566
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Extremely high transformation rates due to innovative electronics and buffers
  • Integrated cuvette chamber
  • Easy to use
  • Compact design
  • Universal electroporator for eukaryotic, yeast and bacteria
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Micro Fusion Chamber
1 Ea.
1
0.5 mm