Learn More
Eppendorf™ Micro Fusion Chamber
Easy to use. Integrated cuvette chamber and extremely high transformation rates
£360.50 - £363.59
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | |||||
15204566
|
Eppendorf™
4308031000 |
0.5 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12816532
|
Eppendorf™
4308030003 |
0.2 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Extremely high transformation rates due to innovative electronics and buffers
- Integrated cuvette chamber
- Easy to use
- Compact design
- Universal electroporator for eukaryotic, yeast and bacteria
Specifications
Micro Fusion Chamber | |
1 Ea. | |
1 |