Learn More
IBA Lifesciences MHC I-Strep HLA-A 2402; SARS-CoV-2 (NYNYLYRLF); Fused to Twin-Strep-tag
A MHC class I molecule fused to a Twin-Strep-tag™ and refolded with a peptide.
Brand: IBA Lifesciences 6-7129-001
Additional Details : Weight : 0.00100kg
Description
- A recombinantly expressed MHC class I molecule that presents a peptide from severe acute respiratory syndrome coronavirus type 2 (SARS-CoV-2). IBA's MHC I Streptamer™ approach is based on IBA's Strep-tag™ technology and facilitates reversible antigen-specific T cell labeling. A Twin-Strep-tag™ fused to the MHC I-Strep permits multimerization of MHC I complexes on a Strep-Tactin™ backbone, resulting in the formation of MHC I Streptamers.
- For multimerizing MHC I-Streps, Strep-Tactin™ TACS Agarose columns, Strep-Tactin™ Magnetic Microbeads or fluorophore-conjugated Strep-Tactin™ can be used. This allows affinity chromatographic or magnetic isolation or cell staining for flow cytometry analysis and sorting (FACS), depending on the experimental requirements. The addition of biotin reverses the multimerization of MHC class I molecules, leading to their dissociation from the target cells. The resulting label-free cells are highly viable and fully functional.
Specifications
10 μg | |
Liquid | |
Antigen-Specific T Cell Labeling | |
108 total cells |