Learn More
IBA Lifesciences MHC I-Strep HLA-A*0201; EBV BMLF1 (GLCTLVAML)
Shop All IBA Lifesciences Products
Click to view available options
:
Capacity: 40μL
Description
Specifications
Specifications
| Capacity (Metric) | 40 μL |
| Kit Contents | Fluorescent reagent for staining, magnetic T cell labeling and isolation |