missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGMT Antibody (MT 23.2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 18 publications
£136.00 - £397.00
Specifications
| Antigen | MGMT |
|---|---|
| Clone | MT 23.2 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:1000, Simple Western 1:1000, Flow Cytometry 1-2 ug/mL, Immunohistochemistry 1:500-1000, Immunocytochemistry/Immunofluorescence 1:500, Immunohistochemistry-Paraffin 1:500-1:1000, Flow (Intracellular), CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18060666
|
Novus Biologicals
NB100-168SS |
0.025 mL |
£136.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18750404
|
Novus Biologicals
NB100-168 |
0.1 mL |
£397.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MGMT Monoclonal specifically detects MGMT in Human, Mouse (Negative) samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular), CyTOF-ready.Specifications
| MGMT | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Chromatin Research, Direct Reversal of DNA Damage, DNA Repair, Tumor Suppressors | |
| PBS with 0.02% Sodium Azide | |
| 6-O-methylguanine-DNA methyltransferase, EC 2.1.1.63, methylated-DNA--protein-cysteine methyltransferase, methylguanine-DNA methyltransferase, O-6-methylguanine-DNA methyltransferase, O6-methylguanine-DNA methyltransferase, O-6-methylguanine-DNA-alkyltransferase | |
| MGMT | |
| IgG2b | |
| Protein G purified | |
| This is specific for MGMT (ref 1). Human. This antibody does not work in mouse. |
| MT 23.2 | |
| Western Blot 1:1000, Simple Western 1:1000, Flow Cytometry 1-2 ug/mL, Immunohistochemistry 1:500-1000, Immunocytochemistry/Immunofluorescence 1:500, Immunohistochemistry-Paraffin 1:500-1:1000, Flow (Intracellular), CyTOF-ready | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human, Mouse (Negative) | |
| P16455 | |
| 4255 | |
| Recombinant human MGMT (O6-methylguanine-DNA methyltransferase) purified from E. coli [UniProt# P16455] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 24 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title