missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGMT Antibody (MT 23.2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 18 publications
Brand: Novus Biologicals NB100-168SS
This item is not returnable.
View return policy
Description
MGMT Monoclonal specifically detects MGMT in Human, Mouse (Negative) samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular), CyTOF-ready.
Specifications
| MGMT | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:1000, Simple Western 1:1000, Flow Cytometry 1-2 ug/mL, Immunohistochemistry 1:500-1000, Immunocytochemistry/Immunofluorescence 1:500, Immunohistochemistry-Paraffin 1:500-1:1000, Flow (Intracellular), CyTOF-ready | |
| P16455 | |
| MGMT | |
| Recombinant human MGMT (O6-methylguanine-DNA methyltransferase) purified from E. coli [UniProt# P16455] | |
| Protein G purified | |
| RUO | |
| Primary | |
| This is specific for MGMT (ref 1). Human. This antibody does not work in mouse. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| MT 23.2 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| 6-O-methylguanine-DNA methyltransferase, EC 2.1.1.63, methylated-DNA--protein-cysteine methyltransferase, methylguanine-DNA methyltransferase, O-6-methylguanine-DNA methyltransferase, O6-methylguanine-DNA methyltransferase, O-6-methylguanine-DNA-alkyltransferase | |
| Mouse | |
| 24 kDa | |
| 0.025 mL | |
| Apoptosis, Cancer, Chromatin Research, Direct Reversal of DNA Damage, DNA Repair, Tumor Suppressors | |
| 4255 | |
| Human, Mouse (Negative) | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction