missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mGluR5/1 Antibody (S75-3), Novus Biologicals™
Mouse Monoclonal Antibody
£168.00
Specifications
| Antigen | metabotropic Glutamate Receptor 5/1 |
|---|---|
| Clone | S75-3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1μg/mL, Immunohistochemistry 0.1-1.0μg/mL, Immunocytochemistry/Immunofluorescence 0.1-1.0μg/mL, Immunoprecipitation 1:10-1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Description
mGluR5/1 Monoclonal specifically detects mGluR5/1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| metabotropic Glutamate Receptor 5/1 | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| glutamate receptor, metabotropic 5, GPRC1EMGLUR5metabotropic glutamate receptor 5, mGlu5, mGluR5 | |
| GRM5 | |
| IgG2a | |
| Protein G purified | |
| Detects approx 130kDa (both mGluR1 and mGluR5). |
| S75-3 | |
| Western Blot 1μg/mL, Immunohistochemistry 0.1-1.0μg/mL, Immunocytochemistry/Immunofluorescence 0.1-1.0μg/mL, Immunoprecipitation 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| P31424 | |
| 2915 | |
| Fusion protein amino acids 824-1203 (cytoplasmic C-terminus) of rat mGluR5b | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title