missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mGluR5/1 Antibody (S75-3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22424-0.025mg
This item is not returnable.
View return policy
Description
mGluR5/1 Monoclonal specifically detects mGluR5/1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| metabotropic Glutamate Receptor 5/1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1μg/mL, Immunohistochemistry 0.1-1.0μg/mL, Immunocytochemistry/Immunofluorescence 0.1-1.0μg/mL, Immunoprecipitation 1:10-1:500 | |
| glutamate receptor, metabotropic 5, GPRC1EMGLUR5metabotropic glutamate receptor 5, mGlu5, mGluR5 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 2915 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| S75-3 | |
| Unconjugated | |
| P31424 | |
| GRM5 | |
| Fusion protein amino acids 824-1203 (cytoplasmic C-terminus) of rat mGluR5b | |
| 0.025 mg | |
| Primary | |
| Detects approx 130kDa (both mGluR1 and mGluR5). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction