missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGAT4A Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | MGAT4A |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18302476
|
Novus Biologicals
NBP3-04923-20UL |
20 μL |
£151.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18360467
|
Novus Biologicals
NBP3-04923-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MGAT4A Polyclonal specifically detects MGAT4A in Human samples. It is validated for Western Blot.Specifications
| MGAT4A | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| alpha-1,3-mannosyl-glycoprotein 4-beta-N-acetylglucosaminyltransferase A, alpha-1,3-mannosyl-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, EC 2.4.1.145, GlcNAc-T IVa, GNT-IV, GNT-IVA, isoenzyme A, isozyme A, mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, N-acetylglucosaminyltransferase IVa, N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase IVa, UDP-GlcNAc:a-1,3-D-mannoside b-1,4-acetylglucosaminyltransferase IV, UDP-N-acetylglucosamine: alpha-1,3-D-mannosidebeta-1,4-N-acetylglucosaminyltransferase IVa, UDP-N-acetylglucosamine:alpha1,3-d-mannosidebeta1,4-N-acetylglucosaminyltransferase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 28-100 of human MGAT4A (NP_036346.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Golgi Apparatus Markers | |
| PBS with 50% glycerol, pH7.3. | |
| 11320 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title