missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius MGA Grade Glass Microfiber Filters w/o Binder
Binder free glass microfiber filters are suitable for analytical and gravimetric analyses. They are also used as prefilter.
£85.70 - £134.00
Specifications
Air Resistance | 36mbar |
---|---|
Color | White |
Format | Disc |
Material | Borosilicate Glass Microfiber |
Particle Retention | 1.6 μm |
Product Code | Brand | Diameter (English) | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) | Diameter (Metric) | Price | Quantity & Availability | ||||
10203431
|
Sartorius
FT-3-1101-125 |
4.92 in. | 12.5 cm |
£106.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
11770416
|
Sartorius
FT-3-1101-185 |
7.2 in. | 18.5 cm |
£85.70
Pack of 50 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
10737271
|
Sartorius
FT-3-1101-150 |
5.9 in. | 15 cm |
£134.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
Description
Applications include: Clarification of buffer & reagent solutions, Filtration of algae cells.Specifications
36mbar | |
Disc | |
1.6 μm | |
40 sec. | |
MGA | |
500°C | |
0.25 mm |
White | |
Borosilicate Glass Microfiber | |
Non-sterile | |
Clarification of buffer and reagent solutions | |
932°F | |
0.009 in. |