missing translation for 'onlineSavingsMsg'
Learn More

Sartorius MGA Grade Glass Microfiber Filters w/o Binder

Binder free glass microfiber filters are suitable for analytical and gravimetric analyses. They are also used as prefilter.

Brand:  Sartorius FT-3-1101-185

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11770416

  • £85.70 / Pack of 50
Estimated Shipment: 30-05-2024
to see stock.
Add to basket

Description

Description

Applications include: Clarification of buffer & reagent solutions, Filtration of algae cells.
Specifications

Specifications

36mbar
Disc
1.6 μm
40 sec.
MGA
500°C
0.25 mm
18.5 cm
White
Borosilicate Glass Microfiber
Non-sterile
Clarification of buffer and reagent solutions
932°F
0.009 in.
7.2 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers