missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MFSD2 Mouse anti-Human, Mouse, Clone: 2D2B4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67965-1-IG-150UL
This item is not returnable.
View return policy
Description
MFSD2 Monoclonal antibody specifically detects MFSD2 in Human, Mouse samples. It is validated for IF, WB, ELISASpecifications
| MFSD2 | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| Mouse | |
| 150 μL | |
| 84879 | |
| Store at -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, ELISA | |
| 2D2B4 | |
| Unconjugated | |
| NM_001031717 | |
| Protein A purification | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction