missing translation for 'onlineSavingsMsg'
Learn More

MFSD2 Mouse anti-Human, Mouse, Clone: 2D2B4, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen MFSD2
Clone 2D2B4
Concentration 1000 μg/mL
Applications Immunofluorescence, Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17701139
View Documents
Proteintech
67965-1-IG-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17791129
View Documents
Proteintech
67965-1-IG-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MFSD2 Monoclonal antibody specifically detects MFSD2 in Human, Mouse samples. It is validated for IF, WB, ELISA
Specifications

Specifications

MFSD2
1000 μg/mL
Monoclonal
Liquid
Human, Mouse
84879
Primary
Store at -20°C
2D2B4
Immunofluorescence, Western Blot, ELISA
Unconjugated
Mouse
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
IgG2a
Protein A purification
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.