missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MFN1 Recombinant Rabbit Monoclonal Antibody (24GB4970)
Rabbit Recombinant Monoclonal Antibody
£172.00 - £488.00
Specifications
| Antigen | MFN1 |
|---|---|
| Clone | 24GB4970 |
| Concentration | 0.7 mg/mL |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30275289
|
Invitrogen™
MA557478 |
100 μL |
£488.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30274812
|
Invitrogen™
MA557479 |
20 μL |
£172.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MFN1 Recombinant Monoclonal Antibody for Western Blot, Flow
Mitofusin 1 (MFN1) and the related protein MFN2 are mitochondrial membrane GTPase proteins that play a central role in mitochondrial metabolism and may be associated with obesity and/or apoptosis processes. MFN1 and MFN2 form homotypic and heterotypic complexes and coordinately regulate mitochondrial fusion and are essential for embryonic development. When ectopically expressed, MFN1 inhibits the apoptosis-associated amino-terminal conformation change in the apoptotic protein Bax but not its mitochondrial translocation, indicating that MFN1 is involved in the regulating the activation of Bax on the outer mitochondrial membrane.Specifications
| MFN1 | |
| 0.7 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q811U4, Q8IWA4, Q8R4Z9 | |
| 192647, 55669, 67414 | |
| A synthesized peptide derived from Rat Mitofusin 1 (aa 116-135). | |
| Antibody |
| 24GB4970 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| MFN1 | |
| 2310002F04Rik; 6330416C07Rik; D3Ertd265e; DKFZp762F247; EC 3.6.5; FLJ20693; Fzo homolog; Fzo1b; hfzo1; hfzo2; HR2; Mfn1; MFN2; MGC41806; mitochondrial mitofusin 1; mitochondrial transmembrane GTPase Fzo-1; mitochondrial transmembrane GTPase FZO1B; mitochondrial transmembrane GTPase FZO-2; mitofusin 1; mitofusin 2; mitofusin-1; mKIAA4032; putative transmembrane GTPase; transmembrane GTPase MFN1 | |
| MFN1 | |
| Primary | |
| Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title